missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Dimethylcyclohexanone, 98%, Thermo Scientific Chemicals
Description
4,4-Dimethylcyclohexanone is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4-Dimethylcyclohexanone is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
4,4-Dimethylcyclohexanone is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Specifications
Specifications
| Melting Point | 41°C to 43°C |
| Flash Point | 62°C (144°F) |
| Quantity | 1 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 126.2 |
| Percent Purity | 98% |
| Chemical Name or Material | 4,4-Dimethylcyclohexanone |
Safety and Handling
| Hazard Category | H228-H319-H500 |
| Precautionary Statement | P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q |
| DOTInformation | Hazard Class: 4.1; Packaging Group: II |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only