missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethylbenzoin 98.0+%, TCI America™

Catalog No. D18815G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D18815G Supplier TCI America Supplier No. D18815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1218-89-9
Molecular Formula C16H16O2
Molecular Weight (g/mol) 240.302
MDL Number MFCD00008555
InChI Key NBYSFWKNMLCZLO-UHFFFAOYSA-N
PubChem CID 224848
IUPAC Name 2-hydroxy-1,2-bis(4-methylphenyl)ethanone
SMILES CC1=CC=C(C=C1)C(C(=O)C2=CC=C(C=C2)C)O

Specifications

Melting Point 88°C
Color White-Yellow
Boiling Point 210°C
Quantity 5 g
Formula Weight 240.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethylbenzoin