accessibility menu, dialog, popup

UserName

CAS RN 1218-89-9

IUPAC Name: 2-hydroxy-1,2-bis(4-methylphenyl)ethan-1-one
Synonyms: 2-hydroxy-1,2-bis(4-methylphenyl)ethanone
Molecular Weight (g/mol): 240.3
Molecular Formula: C16H16O2
InChi Key: NBYSFWKNMLCZLO-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results