missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Dimethylaminophenylazo)phenylarsonic Acid Hydrochloride 95.0+%, TCI America™

Catalog No. D065925G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D065925G 25 g
D06591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D065925G Supplier TCI America Supplier No. D065925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 73688-85-4
Molecular Formula C14H17AsClN3O3
Molecular Weight (g/mol) 385.68
MDL Number MFCD00012589
InChI Key PVYGBUNBLMIFRV-UHFFFAOYSA-N
Synonym 4-(4-Dimethylaminophenylazo)benzenearsonic Acid Hydrochloride
PubChem CID 3056312
IUPAC Name [4-[[4-(dimethylamino)phenyl]diazenyl]phenyl]arsonic acid;hydrochloride
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)[As](=O)(O)O.Cl

Specifications

Color Purple-Red
Quantity 25 g
UN Number 3465
Formula Weight 385.68
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Dimethylaminophenylazo)phenylarsonic Acid Hydrochloride
RTECSNumber CY4800000
TSCA No