missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-(4-Dimethylaminophenylazo)benzoic acid sodium salt, 97%

CAS: 845-46-5 | C15H14N3NaO2 | 291.27 g/mol

Supplier:  Thermo Scientific Chemicals 408110100

 View more versions of this product

Catalog No. AC408110100

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(4-Dimethylaminophenylazo)benzoic acid sodium salt
845-46-5
100.0
Authentic
Glass bottle
MFCD00020350
p-p-dimethylaminophenylazo benzoic acid sodium salt, 4-4-dimethylaminophenylazo benzoic acid sodium salt, benzoic acid, 4-4-dimethylamino phenyl azo-, sodium salt, benzoic acid, p-p-dimethylamino phenyl azo-, sodium salt, benzoic acid, 4-2-4-dimethylamino phenyl diazenyl-, sodium salt 1:1, sodium 4-4-dimethylaminophenylazo benzoate, 4-e-4-dimethylaminophenyl azo benzoic acid
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)[O-].[Na+]
291.27
291.27
Extra Pure
98%
97.5
Orange
97.5% min
C15H14N3NaO2
10 g
OSCKRHPYZNTEIO-UHFFFAOYSA-M
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzoate
23674498
98%
Crystalline Powder

Chemical Identifiers

845-46-5
291.27
OSCKRHPYZNTEIO-UHFFFAOYSA-M
23674498
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)[O-].[Na+]
C15H14N3NaO2
MFCD00020350
p-p-dimethylaminophenylazo benzoic acid sodium salt, 4-4-dimethylaminophenylazo benzoic acid sodium salt, benzoic acid, 4-4-dimethylamino phenyl azo-, sodium salt, benzoic acid, p-p-dimethylamino phenyl azo-, sodium salt, benzoic acid, 4-2-4-dimethylamino phenyl diazenyl-, sodium salt 1:1, sodium 4-4-dimethylaminophenylazo benzoate, 4-e-4-dimethylaminophenyl azo benzoic acid
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only