missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(Dimethylamino)phenylazo]benzoic acid N-succinimidyl ester, 98+%

Catalog No. p-7036153
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54402MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH54402MC Supplier Thermo Scientific Chemicals Supplier No. H54402MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 146998-31-4 | C19H18N4O4 | 366.377 g/mol

4-[4-(Dimethylamino)phenylazo]benzoic acid N-succinimidyl ester is a useful reagent (similar to MANCYL-SE) for labeling proteins or peptides through their amino-groups by forming stable peptide bonds. The succinimidyl ester is reactive with terminal amines or lysines of peptides and other nucleophiles for fluorescent studies of proteins. Its fluorescent properties (has a characteristic broad and intense visible absorption but has no fluorescence) make it an ideal long wavelength quencher and it has been utilized as an acceptor chromophore in FRET studies. It is also used for N-terminal modification of peptides in automated synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-[4-(Dimethylamino)phenylazo]benzoic acid N-succinimidyl ester is a useful reagent (similar to MANCYL-SE) for labeling proteins or peptides through their amino-groups by forming stable peptide bonds. The succinimidyl ester is reactive with terminal amines or lysines of peptides and other nucleophiles for fluorescent studies of proteins. Its fluorescent properties (has a characteristic broad and intense visible absorption but has no fluorescence) make it an ideal long wavelength quencher and it has been utilized as an acceptor chromophore in FRET studies. It is also used for N-terminal modification of peptides in automated synthesis.

Solubility
Soluble in DMSO.

Notes
Moisture sensitive. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 146998-31-4
Molecular Formula C19H18N4O4
Molecular Weight (g/mol) 366.377
MDL Number MFCD00467415
InChI Key IBOVDNBDQHYNJI-UHFFFAOYSA-N
Synonym 2,5-dioxopyrrolidin-1-yl 4-4-dimethylamino phenyl diazenyl benzoate, dabcyl se dye, dabcyl acid, se, 4-4-dimethylamino phenyl azo benzoic acid, succinimidyl ester, 4-4-dimethylamino phenylazo benzoic acid n-succinimidyl ester, dabcyl-nhs, e-2,5-dioxopyrrolidin-1-yl 4-4-dimethylamino phenyl diazenyl benzoate, 2,5-pyrrolidinedione, 1-4-4-dimethylamino phenyl azo benzoyl oxy, dabcyl-se; 4-4-dimethylamino phenylazo benzoic acid n-succinimidyl ester, dabcyl se, solid
PubChem CID 4206604
ChEBI CHEBI:52005
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)ON3C(=O)CCC3=O

Specifications

Quantity 100 mg
Sensitivity Moisture sensitive
Solubility Information Soluble in DMSO.
Formula Weight 366.37
Percent Purity ≥98%
Chemical Name or Material 4-[4-(Dimethylamino)phenylazo]benzoic acid N-succinimidyl ester
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only