accessibility menu, dialog, popup

UserName

CAS RN 146998-31-4

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoate
Synonyms: dabcyl SE dye
Molecular Weight (g/mol): 366.38
Molecular Formula: C19H18N4O4
InChi Key: IBOVDNBDQHYNJI-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)C(=O)ON1C(=O)CCC1=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)