missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-3-thiosemicarbazide 98.0+%, TCI America™

Catalog No. p-7127105
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20961G 1 g
D20965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20961G Supplier TCI America Supplier No. D20961G

Chemical Identifiers

CAS 6926-58-5
Molecular Formula C3H9N3S
Molecular Weight (g/mol) 119.19
MDL Number MFCD00041308
InChI Key FCPHVJQWZFNNKD-UHFFFAOYSA-N
Synonym 4,4-dimethyl-3-thiosemicarbazide, 4,4-dimethylthiosemicarbazide, hydrazinecarbothioamide, n,n-dimethyl, acmc-1b8ou, 1-amino-3,3-dimethylthiourea, 4,4-dimethyl-thiosemicarbazide, n,n-dimethylhydrazinecarbothioamide, hydrazinecarbothioamide,n,n-dimethyl, n,n-dimethylhydrazinecarbothioamide #, dimethylamino hydrazinomethane-1-thione
PubChem CID 2733700
IUPAC Name 3-amino-1,1-dimethylthiourea
SMILES CN(C)C(=S)NN

Specifications

Color White
Quantity 1 g
UN Number 2811
Formula Weight 119.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4-Dimethyl-3-thiosemicarbazide
TSCA No
Recommended Storage Refrigerator