accessibility menu, dialog, popup

UserName

CH3NCH3SNHH2N

CAS RN 6926-58-5

IUPAC Name: 3-amino-1,1-dimethylthiourea
Synonyms: 4,4-dimethylthiosemicarbazide
Molecular Weight (g/mol): 119.19
Molecular Formula: C3H9N3S
InChi Key: FCPHVJQWZFNNKD-UHFFFAOYSA-N
SMILES: CN(C)C(=S)NN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results