missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Dimethyl-3-thiosemicarbazide 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| UN Number | 2811 |
| Formula Weight | 119.19 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,4-Dimethyl-3-thiosemicarbazide |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |