missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane 95.0+%, TCI America™
$54.06 - $192.00
Chemical Identifiers
| CAS | 57280-22-5 |
|---|---|
| Molecular Formula | C7H12O3 |
| Molecular Weight (g/mol) | 144.17 |
| MDL Number | MFCD16621162 |
| InChI Key | GEKNCWQQNMEIMS-UHFFFAOYNA-N |
| PubChem CID | 11094747 |
| IUPAC Name | 4,4-dimethyl-3,5,8-trioxabicyclo[5.1.0]octane |
| SMILES | CC1(C)OCC2OC2CO1 |
Chemical Identifiers
| 57280-22-5 | |
| 144.17 | |
| GEKNCWQQNMEIMS-UHFFFAOYNA-N | |
| 4,4-dimethyl-3,5,8-trioxabicyclo[5.1.0]octane |
| C7H12O3 | |
| MFCD16621162 | |
| 11094747 | |
| CC1(C)OCC2OC2CO1 |
Specifications
| 57280-22-5 | |
| 86°C | |
| MFCD16621162 | |
| GEKNCWQQNMEIMS-UHFFFAOYNA-N | |
| 4,4-dimethyl-3,5,8-trioxabicyclo[5.1.0]octane | |
| 11094747 | |
| ≥95.0% (GC) | |
| 4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane |
| Yellow | |
| C7H12O3 | |
| 1 g | |
| CC1(C)OCC2OC2CO1 | |
| 144.17 | |
| 144.17 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator