accessibility menu, dialog, popup

UserName

CH3OOOCH3

CAS RN 57280-22-5

IUPAC Name: 4,4-dimethyl-3,5,8-trioxabicyclo[5.1.0]octane
Synonyms: 4,4-dimethyl-3,5,8-trioxabicyclo[5.1.0]octane
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: GEKNCWQQNMEIMS-UHFFFAOYNA-N
SMILES: CC1(C)OCC2OC2CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results