missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane 95.0+%, TCI America™

Catalog No. D52005G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52005G 5 g
D52001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52005G Supplier TCI America Supplier No. D52005G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57280-22-5
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD16621162
InChI Key GEKNCWQQNMEIMS-UHFFFAOYNA-N
PubChem CID 11094747
IUPAC Name 4,4-dimethyl-3,5,8-trioxabicyclo[5.1.0]octane
SMILES CC1(C)OCC2OC2CO1

Specifications

Color Yellow
Boiling Point 86°C
Quantity 5 g
Formula Weight 144.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane
TSCA No
Recommended Storage Refrigerator