Learn More
4,4-Dimethyl-2-pyrrolidinone, 95%
CAS: 66899-02-3 | C6H11NO | 113.16 g/mol
Supplier: Thermo Scientific Chemicals H3320903
Description
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications
| 4,4-Dimethyl-2-pyrrolidinone | |
| 38°C to 40°C | |
| C6H11NO | |
| 1 g | |
| 4,4-dimethyl-2-pyrrolidinone, 2-pyrrolidinone, 4,4-dimethyl, 4,4-dimethylpyrrolidine-2-one, 2-pyrrolidone,4,4-dimethyl, 4,4-dimethyl-2-pyrrolidone, 4,4-dimethyl-pyrrolidin-2-one, 4, 4-dimethyl-2-pyrrolidinone | |
| HGVPKAGCVCGRDQ-UHFFFAOYSA-N | |
| 4,4-dimethylpyrrolidin-2-one | |
| 637593 | |
| 95% |
| 66899-02-3 | |
| White to Cream | |
| MFCD09864496 | |
| Air Sensitive | |
| Sparingly soluble in water.(0.26g/L) (25°C), | |
| CC1(CC(=O)NC1)C | |
| 113.16 | |
| 113.2 | |
| Solid |
Chemical Identifiers
| 66899-02-3 | |
| 113.16 | |
| HGVPKAGCVCGRDQ-UHFFFAOYSA-N | |
| 4,4-dimethylpyrrolidin-2-one |
| C6H11NO | |
| MFCD09864496 | |
| 637593 | |
| CC1(CC(=O)NC1)C |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only