accessibility menu, dialog, popup

UserName

CAS RN 66899-02-3

H3COHNCH3

CAS RN 66899-02-3

IUPAC Name: 4,4-dimethylpyrrolidin-2-one
Synonyms: 4,4-dimethylpyrrolidin-2-one
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: HGVPKAGCVCGRDQ-UHFFFAOYSA-N
SMILES: CC1(C)CNC(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results