missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-2-pyrrolidinone, 95%

Catalog No. AAH33209MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33209MD 250 mg
AAH3320903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33209MD Supplier Thermo Scientific Chemicals Supplier No. H33209MD

CAS: 66899-02-3 | C6H11NO | 113.16 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 66899-02-3
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD09864496
InChI Key HGVPKAGCVCGRDQ-UHFFFAOYSA-N
PubChem CID 637593
IUPAC Name 4,4-dimethylpyrrolidin-2-one
SMILES CC1(CC(=O)NC1)C

Specifications

Melting Point 38°C to 40°C
Color White to Cream
Quantity 250 mg
Sensitivity Air Sensitive
Synonym 4,4-dimethyl-2-pyrrolidinone, 2-pyrrolidinone, 4,4-dimethyl, 4,4-dimethylpyrrolidine-2-one, 2-pyrrolidone,4,4-dimethyl, 4,4-dimethyl-2-pyrrolidone, 4,4-dimethyl-pyrrolidin-2-one, 4, 4-dimethyl-2-pyrrolidinone
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 113.2
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4,4-Dimethyl-2-pyrrolidinone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only