missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-2-cyclohexen-1-one, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1073-13-8
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.18
MDL Number MFCD00009695
InChI Key HAUNPYVLVAIUOO-UHFFFAOYSA-N
Synonym 4,4-dimethyl-2-cyclohexen-1-one, 4,4-dimethylcyclohex-2-enone, 2-cyclohexen-1-one, 4,4-dimethyl, 4,4-dimethyl-2-cyclohexenone, 4,4-dimethylcyclohexenone, 4,4-dimethyl-2-cyclohexene-1-one, 4,4-dimethyl-cyclohex-2-en-1-one, pubchem17152, acmc-1bxzo, 3,3-dimethylcyclohexene-6-one
PubChem CID 136839
IUPAC Name 4,4-dimethylcyclohex-2-en-1-one
SMILES CC1(C)CCC(=O)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC319540050
SDS View Documents
Thermo Scientific Chemicals
319540050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1073-13-8
124.18
HAUNPYVLVAIUOO-UHFFFAOYSA-N
136839
CC1(C)CCC(=O)C=C1
C8H12O
MFCD00009695
4,4-dimethyl-2-cyclohexen-1-one, 4,4-dimethylcyclohex-2-enone, 2-cyclohexen-1-one, 4,4-dimethyl, 4,4-dimethyl-2-cyclohexenone, 4,4-dimethylcyclohexenone, 4,4-dimethyl-2-cyclohexene-1-one, 4,4-dimethyl-cyclohex-2-en-1-one, pubchem17152, acmc-1bxzo, 3,3-dimethylcyclohexene-6-one
4,4-dimethylcyclohex-2-en-1-one

Specifications

1073-13-8
100.0
72.0°C to 73.5°C (20.0mmHg)
Authentic
Glass bottle
1.4710 to 1.4740
MFCD00009695
0.94
HAUNPYVLVAIUOO-UHFFFAOYSA-N
4,4-dimethylcyclohex-2-en-1-one
136839
97%
96.0
0.9400g/mL
64°C
96% min. (GC)
C8H12O
(CH3)2C6H6(=O)
5g
4,4-dimethyl-2-cyclohexen-1-one, 4,4-dimethylcyclohex-2-enone, 2-cyclohexen-1-one, 4,4-dimethyl, 4,4-dimethyl-2-cyclohexenone, 4,4-dimethylcyclohexenone, 4,4-dimethyl-2-cyclohexene-1-one, 4,4-dimethyl-cyclohex-2-en-1-one, pubchem17152, acmc-1bxzo, 3,3-dimethylcyclohexene-6-one
CC1(C)CCC(=O)C=C1
124.18
124.18
4, 4-Dimethyl-2-cyclohexen-1-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only