accessibility menu, dialog, popup

UserName

CH3H3CO

CAS RN 1073-13-8

IUPAC Name: 4,4-dimethylcyclohex-2-en-1-one
Synonyms: 4,4-dimethylcyclohex-2-en-1-one
Molecular Weight (g/mol): 124.18
Molecular Formula: C8H12O
InChi Key: HAUNPYVLVAIUOO-UHFFFAOYSA-N
SMILES: CC1(C)CCC(=O)C=C1
Boiling Point 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)