missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-2-cyclohexen-1-one-2-boronic acid pinacol ester, 97%

Catalog No. AC444980010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444980010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC444980010 Supplier Thermo Scientific Chemicals Supplier No. 444980010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 219489-09-5 | C14H23BO3 | 250.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 219489-09-5
Molecular Formula C14H23BO3
Molecular Weight (g/mol) 250.15
MDL Number MFCD09027074
InChI Key MMYLPQABFZSMCP-UHFFFAOYSA-N
Synonym 4,4-dimethylcyclohex-2-en-1-one-2-boronic acid, pinacol ester, 4,4-dimethyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohex-2-en-1-one, 4,4-dimethyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohex-2-enone, 4,4-dimethylcyclohex-2-enone-2-boronic acid, pinacol ester, 4,4-dimethylcyclohex-2-en-1-one-2-boronic acid pinacol ester, 4,4-dimethyl-2-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohex-2-en-1-one, 4,4-dimethylcyclohex-2-enone-2-boronic acid pinacol ester, 4,4-dimethyl cyclohex-2-en-1-one-2-boronic acid pinacol ester, 2-3,3-dimethyl-6-oxocyclohex-1-en-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 10220580
IUPAC Name 4,4-dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-2-en-1-one
SMILES CC1(C)OB(OC1(C)C)C1=CC(C)(C)CCC1=O

Specifications

Melting Point 99°C to 102°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 250.15
Percent Purity 97%
Chemical Name or Material 4,4-Dimethyl-2-cyclohexen-1-one-2-boronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only