accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3COCH3CH3

CAS RN 219489-09-5

IUPAC Name: 4,4-dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-2-en-1-one
Synonyms: 4,4-dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-2-en-1-one
Molecular Weight (g/mol): 250.15
Molecular Formula: C14H23BO3
InChi Key: MMYLPQABFZSMCP-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(C)(C)CCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results