missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-1-phenylpentane-1,3-dione, 99%

Catalog No. AAL1525106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1525106 5 g
AAL1525114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1525106 Supplier Thermo Scientific Chemicals Supplier No. L1525106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13988-67-5 | C13H16O2 | 204.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13988-67-5
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.27
MDL Number MFCD00511275
InChI Key HORVLKADAZQYRS-UHFFFAOYSA-N
Synonym cbmicro_020484, benzoylpivaloylmethane, acmc-1c1ej, 2-tert-butylcarbonyl acetophenone, 1-tert-butyl-3-phenylmalonaldehyde, 1,3-pentanedione,4,4-dimethyl-1-phenyl, 1,3-pentanedione, 4,4-dimethyl-1-phenyl, 4,4-dimethyl-1-phenyl-1,3-pentanedione #
PubChem CID 589971
IUPAC Name 4,4-dimethyl-1-phenylpentane-1,3-dione
SMILES CC(C)(C)C(=O)CC(=O)C1=CC=CC=C1

Specifications

Density 1.024
Boiling Point 127°C to 128°C (2 mmHg)
Refractive Index 1.559
Quantity 5 g
Beilstein 1868436
Formula Weight 204.27
Percent Purity 99%
Chemical Name or Material 4,4-Dimethyl-1-phenylpentane-1,3-dione
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only