CAS RN 13988-67-5
IUPAC Name:
4,4-dimethyl-1-phenylpentane-1,3-dione
Synonyms:
4,4-dimethyl-1-phenylpentane-1,3-dione
Molecular Weight (g/mol):
204.27
Molecular Formula:
C13H16O2
InChi Key:
HORVLKADAZQYRS-UHFFFAOYSA-N
SMILES:
CC(C)(C)C(=O)CC(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
4,4-Dimethyl-1-phenylpentane-1,3-dione, 99%
4,4-Dimethyl-1-phenylpentane-1,3-dione, 99%
CAS: 13988-67-5 Molecular Formula: C13H16O2 Molecular Weight (g/mol): 204.27 MDL Number: MFCD00511275 InChI Key: HORVLKADAZQYRS-UHFFFAOYSA-N Synonym: cbmicro_020484,benzoylpivaloylmethane,acmc-1c1ej,2-tert-butylcarbonyl acetophenone,1-tert-butyl-3-phenylmalonaldehyde,1,3-pentanedione,4,4-dimethyl-1-phenyl,1,3-pentanedione, 4,4-dimethyl-1-phenyl,4,4-dimethyl-1-phenyl-1,3-pentanedione # PubChem CID: 589971 IUPAC Name: 4,4-dimethyl-1-phenylpentane-1,3-dione SMILES: CC(C)(C)C(=O)CC(=O)C1=CC=CC=C1