missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dimethoxytrityl Chloride 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 125°C |
| Quantity | 5 g |
| Formula Weight | 338.83 |
| Percent Purity | ≥97.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,4′-Dimethoxytrityl Chloride [Hydroxyl Protecting Agent] |
Safety and Handling
| EINECSNumber | (4)-1881 |
| TSCA | No |