missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxytrityl chloride, 97%

Catalog No. aaa1162614
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1162614 25 g
AAA1162606 5 g
AAA1162622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1162614 Supplier Thermo Scientific Chemicals Supplier No. A1162614
Only null left

CAS: 40615-36-9 | C21H19ClO2 | 338.83 g/mol

4,4'-Dimethoxytrityl chloride is used as a protective reagent for oligonucleotide synthesis. It is used for selective protection and deprotection procedures for thiol and hydroxy groups in nucleoside derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethoxytrityl chloride is used as a protective reagent for oligonucleotide synthesis. It is used for selective protection and deprotection procedures for thiol and hydroxy groups in nucleoside derivatives.

Solubility
Soluble in chloroform and methanol. Partially soluble in water

Notes
Moisture sensitive, hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 40615-36-9
Molecular Formula C21H19ClO2
Molecular Weight (g/mol) 338.83
MDL Number MFCD00008409
InChI Key JBWYRBLDOOOJEU-UHFFFAOYSA-N
Synonym 4,4'-dimethoxytrityl chloride, dmt-cl, 4,4'-dimethoxytritylchloride, 4,4'-chloro phenyl methylene bis methoxybenzene, 4,4'-dimethoxytriphenylmethyl chloride, 1,1'-chlorophenylmethylene bis 4-methoxybenzene, benzene, 1,1'-chlorophenylmethylene bis 4-methoxy, chloro-4,4'-dimethoxytriphenylmethane, 1-chloro 4-methoxyphenyl phenylmethyl-4-methoxybenzene, 4,4'-dimethoxytritylchloride dmt-cl
PubChem CID 96831
IUPAC Name 1-[chloro-(4-methoxyphenyl)-phenylmethyl]-4-methoxybenzene
SMILES COC1=CC=C(C=C1)C(Cl)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Specifications

Melting Point 121°C to 127°C
Quantity 25 g
Beilstein 2471942
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform and methanol. Partially soluble in water
Formula Weight 338.83
Percent Purity 98%
Chemical Name or Material 4,4'-Dimethoxytrityl chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 255-002-6
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only