missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dimethoxyoctafluorobiphenyl 98.0+%, TCI America™
$51.98 - $157.87
Chemical Identifiers
| CAS | 2200-71-7 |
|---|---|
| Molecular Formula | C14H6F8O2 |
| Molecular Weight (g/mol) | 358.19 |
| MDL Number | MFCD00039802 |
| InChI Key | TVUPJKALGKCABC-UHFFFAOYSA-N |
| PubChem CID | 262954 |
| IUPAC Name | 2,2',3,3',5,5',6,6'-octafluoro-4,4'-dimethoxy-1,1'-biphenyl |
| SMILES | COC1=C(F)C(F)=C(C(F)=C1F)C1=C(F)C(F)=C(OC)C(F)=C1F |
Chemical Identifiers
| 2200-71-7 | |
| 358.19 | |
| TVUPJKALGKCABC-UHFFFAOYSA-N | |
| 2,2',3,3',5,5',6,6'-octafluoro-4,4'-dimethoxy-1,1'-biphenyl |
| C14H6F8O2 | |
| MFCD00039802 | |
| 262954 | |
| COC1=C(F)C(F)=C(C(F)=C1F)C1=C(F)C(F)=C(OC)C(F)=C1F |
Specifications
| 2200-71-7 | |
| White-Yellow | |
| MFCD00039802 | |
| Octafluoro-4,4′C-dimethoxybiphenyl | |
| COC1=C(F)C(F)=C(C(F)=C1F)C1=C(F)C(F)=C(OC)C(F)=C1F | |
| 358.19 | |
| 358.19 | |
| Crystalline Powder |
| 87°C | |
| C14H6F8O2 | |
| 1 g | |
| TVUPJKALGKCABC-UHFFFAOYSA-N | |
| 2,2',3,3',5,5',6,6'-octafluoro-4,4'-dimethoxy-1,1'-biphenyl | |
| 262954 | |
| ≥98.0% (GC) | |
| 4,4′-Dimethoxyoctafluorobiphenyl |
Safety and Handling
TSCA : No