missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxybenzophenone 99.0+%, TCI America™

Catalog No. p-7126417
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D076525G 25 g
D0765100G 100 g
D0765500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D076525G Supplier TCI America Supplier No. D076525G
Only null left

Chemical Identifiers

CAS 90-96-0
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00008404
InChI Key RFVHVYKVRGKLNK-UHFFFAOYSA-N
Synonym 4,4'-dimethoxybenzophenone, bis 4-methoxyphenyl methanone, methanone, bis 4-methoxyphenyl, p,p'-dimethoxybenzophenone, dmbp, bis p-anisyl ketone, bis 4-anisyl ketone, bis p-methoxy benzophenone, benzophenone, 4,4'-dimethoxy, 1,1-di p-methoxyphenyl-ketone
PubChem CID 7032
IUPAC Name bis(4-methoxyphenyl)methanone
SMILES COC1=CC=C(C=C1)C(=O)C1=CC=C(OC)C=C1

Specifications

Melting Point 145°C
Color White
Quantity 25 g
Formula Weight 242.27
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethoxybenzophenone
EINECSNumber (4)-1467
TSCA No