missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dimethoxy-2,2'-bipyridyl 98.0+%, TCI America™
$119.30 - $369.08
Chemical Identifiers
| CAS | 17217-57-1 |
|---|---|
| Molecular Formula | C12H12N2O2 |
| Molecular Weight (g/mol) | 216.24 |
| MDL Number | MFCD00233880 |
| InChI Key | IMEVSAIFJKKDAP-UHFFFAOYSA-N |
| Synonym | 4,4'-dimethoxy-2,2'-bipyridine, 4,4'-dimethoxy-2,2'-bipyridyl, 4-methoxy-2-4-methoxypyridin-2-yl pyridine, 4,4'-dimethoxy-2,2' bipyridinyl, 2,2'-bipyridine, 4,4'-dimethoxy, acmc-1c6x1, 4-4'-dimethoxy-2-2'-bipyridine, 4-methoxy-2-4-methoxy 2-pyridyl pyridine, 4-methoxy-2-4-methoxy-2-pyridinyl pyridine |
| PubChem CID | 2733927 |
| IUPAC Name | 4,4'-dimethoxy-2,2'-bipyridine |
| SMILES | COC1=CC=NC(=C1)C1=CC(OC)=CC=N1 |
Chemical Identifiers
| 17217-57-1 | |
| 216.24 | |
| IMEVSAIFJKKDAP-UHFFFAOYSA-N | |
| 2733927 | |
| COC1=CC=NC(=C1)C1=CC(OC)=CC=N1 |
| C12H12N2O2 | |
| MFCD00233880 | |
| 4,4'-dimethoxy-2,2'-bipyridine, 4,4'-dimethoxy-2,2'-bipyridyl, 4-methoxy-2-4-methoxypyridin-2-yl pyridine, 4,4'-dimethoxy-2,2' bipyridinyl, 2,2'-bipyridine, 4,4'-dimethoxy, acmc-1c6x1, 4-4'-dimethoxy-2-2'-bipyridine, 4-methoxy-2-4-methoxy 2-pyridyl pyridine, 4-methoxy-2-4-methoxy-2-pyridinyl pyridine | |
| 4,4'-dimethoxy-2,2'-bipyridine |
Specifications
| 17217-57-1 | |
| White-Yellow | |
| MFCD00233880 | |
| 4,4'-dimethoxy-2,2'-bipyridine, 4,4'-dimethoxy-2,2'-bipyridyl, 4-methoxy-2-4-methoxypyridin-2-yl pyridine, 4,4'-dimethoxy-2,2' bipyridinyl, 2,2'-bipyridine, 4,4'-dimethoxy, acmc-1c6x1, 4-4'-dimethoxy-2-2'-bipyridine, 4-methoxy-2-4-methoxy 2-pyridyl pyridine, 4-methoxy-2-4-methoxy-2-pyridinyl pyridine | |
| COC1=CC=NC(=C1)C1=CC(OC)=CC=N1 | |
| 216.24 | |
| 216.24 | |
| Crystalline Powder |
| 172°C | |
| C12H12N2O2 | |
| 1 g | |
| IMEVSAIFJKKDAP-UHFFFAOYSA-N | |
| 4,4'-dimethoxy-2,2'-bipyridine | |
| 2733927 | |
| ≥98.0% (GC,T) | |
| 4,4′-Dimethoxy-2,2′-bipyridyl |
Safety and Handling
TSCA : No