accessibility menu, dialog, popup

UserName

CAS RN 17217-57-1

CH3ONOCH3N

CAS RN 17217-57-1

IUPAC Name: 4,4'-dimethoxy-2,2'-bipyridine
Synonyms: 4,4'-dimethoxy-2,2'-bipyridine
Molecular Weight (g/mol): 216.24
Molecular Formula: C12H12N2O2
InChi Key: IMEVSAIFJKKDAP-UHFFFAOYSA-N
SMILES: COC1=CC=NC(=C1)C1=CC(OC)=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results