missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dimethoxy-2,2'-bipyridyl 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 172°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 216.24 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,4′-Dimethoxy-2,2′-bipyridyl |
Safety and Handling
| TSCA | No |