missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxy-2,2'-bipyridyl 98.0+%, TCI America™

Catalog No. D38861G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38861G 1 g
D38865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38861G Supplier TCI America Supplier No. D38861G
Only null left

Chemical Identifiers

CAS 17217-57-1
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
MDL Number MFCD00233880
InChI Key IMEVSAIFJKKDAP-UHFFFAOYSA-N
Synonym 4,4'-dimethoxy-2,2'-bipyridine, 4,4'-dimethoxy-2,2'-bipyridyl, 4-methoxy-2-4-methoxypyridin-2-yl pyridine, 4,4'-dimethoxy-2,2' bipyridinyl, 2,2'-bipyridine, 4,4'-dimethoxy, acmc-1c6x1, 4-4'-dimethoxy-2-2'-bipyridine, 4-methoxy-2-4-methoxy 2-pyridyl pyridine, 4-methoxy-2-4-methoxy-2-pyridinyl pyridine
PubChem CID 2733927
IUPAC Name 4,4'-dimethoxy-2,2'-bipyridine
SMILES COC1=CC=NC(=C1)C1=CC(OC)=CC=N1

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 1 g
Formula Weight 216.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethoxy-2,2′-bipyridyl