missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diiodobiphenyl 98.0+%, TCI America™

Catalog No. p-7127224
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D231525G 25 g
D2315250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D231525G Supplier TCI America Supplier No. D231525G
Only null left

Chemical Identifiers

CAS 3001-15-8
Molecular Formula C12H8I2
Molecular Weight (g/mol) 406.005
MDL Number MFCD00001057
InChI Key GPYDMVZCPRONLW-UHFFFAOYSA-N
Synonym 4,4'-diiodobiphenyl, 4,4'-diiodo-1,1'-biphenyl, 4,4'-diiododiphenyl, 1,1'-biphenyl, 4,4'-diiodo, 1-iodo-4-4-iodophenyl benzene, 4,4-diiodobiphenyl, 4-4'-iodophenyl iodobenzene, 4,4;-diiodobiphenyl, 4,4'-diiododophenyl, pract, 4-iodo-1-4-iodophenyl benzene
PubChem CID 76348
IUPAC Name 1-iodo-4-(4-iodophenyl)benzene
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)I)I

Specifications

Melting Point 208°C
Color Yellow
Quantity 25 g
UN Number 3152
Formula Weight 406.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diiodobiphenyl
EINECSNumber (4)-1507
TSCA Yes