missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diiodo-2,2'-dimethylbiphenyl 98.0+%, TCI America™

Catalog No. D46085G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46085G 5 g
D46081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46085G Supplier TCI America Supplier No. D46085G

Chemical Identifiers

CAS 69571-02-4
Molecular Formula C14H12I2
Molecular Weight (g/mol) 434.06
MDL Number MFCD13185944
InChI Key WHFMNDMHTIOIMS-UHFFFAOYSA-N
PubChem CID 13797298
IUPAC Name 4,4'-diiodo-2,2'-dimethyl-1,1'-biphenyl
SMILES CC1=C(C=CC(I)=C1)C1=C(C)C=C(I)C=C1

Specifications

Melting Point 98°C
Color White
Quantity 5 g
Formula Weight 434.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diiodo-2,2′-dimethylbiphenyl