missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dihydroxybiphenyl 99.0+%, TCI America™

Catalog No. p-7118460
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B046425G 25 g
B0464100G 100 g
B0464500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B046425G Supplier TCI America Supplier No. B046425G
Only null left

Chemical Identifiers

CAS 92-88-6
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00002348
InChI Key VCCBEIPGXKNHFW-UHFFFAOYSA-N
Synonym 4,4'-dihydroxybiphenyl, 4,4'-biphenol, 4,4'-biphenyldiol, biphenyl-4,4'-diol, 1,1'-biphenyl-4,4'-diol, p,p'-biphenol, p,p'-diphenol, antioxidant dod, 4,4'-dioxydiphenyl, p-dihydroxydiphenyl
PubChem CID 7112
ChEBI CHEBI:34367
IUPAC Name 4-(4-hydroxyphenyl)phenol
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)O)O

Specifications

Melting Point 281°C
Quantity 25 g
UN Number 3077
Formula Weight 186.21
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dihydroxybiphenyl
EINECSNumber (4)-0820
RTECSNumber DV4725000
TSCA Yes