missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dihydroxy-3,3',5,5'-tetraisopropylbiphenyl 97.0+%, TCI America™

Catalog No. p-7128724
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5281200MG 200 mg
D52811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5281200MG Supplier TCI America Supplier No. D5281200MG
Only null left

Chemical Identifiers

CAS 2416-95-7
Molecular Formula C24H34O2
Molecular Weight (g/mol) 354.53
MDL Number MFCD20257818
InChI Key QAISRHCMPQROAX-UHFFFAOYSA-N
Synonym 3,3′,5,5′-Tetraisopropyl-[1,1′-biphenyl]-4,4′-diol
PubChem CID 11602828
IUPAC Name 3,3',5,5'-tetrakis(propan-2-yl)-[1,1'-biphenyl]-4,4'-diol
SMILES CC(C)C1=CC(=CC(C(C)C)=C1O)C1=CC(C(C)C)=C(O)C(=C1)C(C)C

Specifications

Melting Point 109°C
Quantity 200 mg
Formula Weight 354.53
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dihydroxy-3,3′,5,5′-tetraisopropylbiphenyl
TSCA No
Recommended Storage Refrigerator