missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Difluorobenzophenone 99.0+%, TCI America™

Catalog No. D1797500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1797500G 500 g
D179725G 25 g
D1797100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1797500G Supplier TCI America Supplier No. D1797500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 345-92-6
Molecular Formula C13H8F2O
Molecular Weight (g/mol) 218.20
MDL Number MFCD00000353
InChI Key LSQARZALBDFYQZ-UHFFFAOYSA-N
Synonym 4,4'-difluorobenzophenone, bis 4-fluorophenyl methanone, bis 4-fluorophenyl-methanone, methanone, bis 4-fluorophenyl, p,p'-difluorobenzophenone, di-p-fluorophenyl ketone, bis p-fluorophenyl ketone, bis 4-fluorophenyl ketone, benzophenone, 4,4'-difluoro, unii-88bnc11b9c
PubChem CID 9582
IUPAC Name bis(4-fluorophenyl)methanone
SMILES FC1=CC=C(C=C1)C(=O)C1=CC=C(F)C=C1

Specifications

Melting Point 108°C
Color White
Quantity 500 g
Formula Weight 218.20
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Difluorobenzophenone
RTECSNumber DJ0685000
TSCA No