missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Difluorobenzhydrol 98.0+%, TCI America™

Catalog No. D179525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D179525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D179525G Supplier TCI America Supplier No. D179525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 365-24-2
Molecular Formula C13H10F2O
Molecular Weight (g/mol) 220.219
MDL Number MFCD00000357
InChI Key WCTZPQWLFWZYJE-UHFFFAOYSA-N
Synonym 4,4'-difluorobenzhydrol, bis 4-fluorophenyl methanol, bis-4-fluorophenyl-methanol, 4,4'-difluorobenzhydryl alcohol, bis 4-fluorophenyl methan-1-ol, 4,4'-diflurobenzylmethanol, pubchem14058, 4,4'-diflourobenzhydrol, 4,4'-difluoro benzhydrol, 4,4'-difluoro-benzhydrol
PubChem CID 123049
IUPAC Name bis(4-fluorophenyl)methanol
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)F)O)F

Specifications

Melting Point 48°C
Boiling Point 154°C
Quantity 25 g
Formula Weight 220.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Difluorobenzhydrol