missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Difluorobenzhydrol, 98+%

Catalog No. aaa1437014
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1437014 25 g
AAA1437006 5 g
AAA1437022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1437014 Supplier Thermo Scientific Chemicals Supplier No. A1437014
Only null left

CAS: 365-24-2 | C13H10F2O | 220.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 365-24-2
Molecular Formula C13H10F2O
Molecular Weight (g/mol) 220.219
MDL Number MFCD00000357
InChI Key WCTZPQWLFWZYJE-UHFFFAOYSA-N
Synonym 4,4'-difluorobenzhydrol, bis 4-fluorophenyl methanol, bis-4-fluorophenyl-methanol, 4,4'-difluorobenzhydryl alcohol, bis 4-fluorophenyl methan-1-ol, 4,4'-diflurobenzylmethanol, pubchem14058, 4,4'-diflourobenzhydrol, 4,4'-difluoro benzhydrol, 4,4'-difluoro-benzhydrol
PubChem CID 123049
IUPAC Name bis(4-fluorophenyl)methanol
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)F)O)F

Specifications

Melting Point 45°C to 49°C
Boiling Point 142°C to 144°C (3 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 2332258
Formula Weight 220.22
Percent Purity ≥98%
Chemical Name or Material 4,4'-Difluorobenzhydrol
EINECSNumber 206-671-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only