missing translation for 'onlineSavingsMsg'
Learn More

4,4-Difluoro-1,3-dimethyl-4-bora-3a,4a-diaza-s-indacene 98.0+%, TCI America™

Catalog No. D4897200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4897200MG 200 mg
D48971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4897200MG Supplier TCI America Supplier No. D4897200MG

Chemical Identifiers

CAS 154793-49-4
Molecular Formula C11H11BF2N2
Molecular Weight (g/mol) 220.03
InChI Key PIHALWZKFOHBCB-UHFFFAOYSA-N
PubChem CID 124201520
SMILES [B-]1(N2C(=CC(=C2C=C3[N+]1=CC=C3)C)C)(F)F

Specifications

Melting Point 138°C
Quantity 200 mg
Formula Weight 220.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4-Difluoro-1,3-dimethyl-4-bora-3a,4a-diaza-s-indacene