accessibility menu, dialog, popup

UserName

BNNCH3CH3FF

CAS RN 154793-49-4

IUPAC Name: 2,2-difluoro-4,6-dimethyl-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide
Synonyms: 2,2-difluoro-4,6-dimethyl-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide
Molecular Weight (g/mol): 220.03
Molecular Formula: C11H11BF2N2
InChi Key: PIHALWZKFOHBCB-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C2C=C3C=CC=[N+]3[B-](F)(F)N12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results