missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diethylbiphenyl 98.0+%, TCI America™

Catalog No. D293825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D293825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D293825G Supplier TCI America Supplier No. D293825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13049-40-6
Molecular Formula C16H18
Molecular Weight (g/mol) 210.32
MDL Number MFCD06797093
InChI Key UMSGIWAAMHRVQI-UHFFFAOYSA-N
PubChem CID 42292
IUPAC Name 1-ethyl-4-(4-ethylphenyl)benzene
SMILES CCC1=CC=C(C=C1)C2=CC=C(C=C2)CC

Specifications

Melting Point 82°C
Color White
Boiling Point 315°C
Quantity 25 g
Formula Weight 210.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diethylbiphenyl
EINECSNumber (4)-0016
TSCA No