accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 13049-40-6

IUPAC Name: 4,4'-diethyl-1,1'-biphenyl
Synonyms: 4,4'-diethyl-1,1'-biphenyl
Molecular Weight (g/mol): 210.32
Molecular Formula: C16H18
InChi Key: UMSGIWAAMHRVQI-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)C1=CC=C(CC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results