missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Diethoxyazobenzene, 97%
CAS: 588-52-3 | C16H18N2O2 | 270.33 g/mol
$54.56 - $178.30
Chemical Identifiers
| CAS | 588-52-3 |
|---|---|
| Molecular Formula | C16H18N2O2 |
| Molecular Weight (g/mol) | 270.33 |
| MDL Number | MFCD00053756 |
| InChI Key | FMPYMMTZQFFROU-UHFFFAOYSA-N |
| Synonym | 4,4'-diethoxyazobenzene, diazene, bis 4-ethoxyphenyl, 4,4'-azodiphenetole, bis 4-ethoxyphenyl diazene, 4,4'-azophenetole, bis-p-ethoxyazobenzene, e-bis 4-ethoxyphenyl diazene, diazene, 1,2-bis 4-ethoxyphenyl, phenetole, 4,4'-azodi-6ci,7ci, acmc-20amss |
| PubChem CID | 101783 |
| IUPAC Name | bis(4-ethoxyphenyl)diazene |
| SMILES | CCOC1=CC=C(C=C1)N=NC1=CC=C(OCC)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2083903
|
Thermo Scientific Chemicals
B2083903 |
1 g |
Each for $54.56
|
|
|||||
|
AAB2083906
|
Thermo Scientific Chemicals
B2083906 |
5 g |
Each for $178.30
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 588-52-3 | |
| 270.33 | |
| FMPYMMTZQFFROU-UHFFFAOYSA-N | |
| 101783 | |
| CCOC1=CC=C(C=C1)N=NC1=CC=C(OCC)C=C1 |
| C16H18N2O2 | |
| MFCD00053756 | |
| 4,4'-diethoxyazobenzene, diazene, bis 4-ethoxyphenyl, 4,4'-azodiphenetole, bis 4-ethoxyphenyl diazene, 4,4'-azophenetole, bis-p-ethoxyazobenzene, e-bis 4-ethoxyphenyl diazene, diazene, 1,2-bis 4-ethoxyphenyl, phenetole, 4,4'-azodi-6ci,7ci, acmc-20amss | |
| bis(4-ethoxyphenyl)diazene |
Specifications
| 588-52-3 | |
| MFCD00053756 | |
| 4,4'-diethoxyazobenzene, diazene, bis 4-ethoxyphenyl, 4,4'-azodiphenetole, bis 4-ethoxyphenyl diazene, 4,4'-azophenetole, bis-p-ethoxyazobenzene, e-bis 4-ethoxyphenyl diazene, diazene, 1,2-bis 4-ethoxyphenyl, phenetole, 4,4'-azodi-6ci,7ci, acmc-20amss | |
| CCOC1=CC=C(C=C1)N=NC1=CC=C(OCC)C=C1 | |
| 270.33 | |
| 270.33 | |
| 4,4'-Diethoxyazobenzene |
| C16H18N2O2 | |
| 1 g | |
| FMPYMMTZQFFROU-UHFFFAOYSA-N | |
| bis(4-ethoxyphenyl)diazene | |
| 101783 | |
| 97% |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
RTECSNumber : HM2966300
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only