CAS RN 588-52-3
IUPAC Name:
bis(4-ethoxyphenyl)diazene
Synonyms:
diazene, bis(4-ethoxyphenyl)-
Molecular Weight (g/mol):
270.33
Molecular Formula:
C16H18N2O2
InChi Key:
FMPYMMTZQFFROU-UHFFFAOYSA-N
SMILES:
CCOC1=CC=C(C=C1)N=NC1=CC=C(OCC)C=C1
1
–
1
of
1
results
1
4,4'-Diethoxyazobenzene, 97%
CAS: 588-52-3 Molecular Formula: C16H18N2O2 Molecular Weight (g/mol): 270.33 MDL Number: MFCD00053756 InChI Key: FMPYMMTZQFFROU-UHFFFAOYSA-N Synonym: 4,4'-diethoxyazobenzene,diazene, bis 4-ethoxyphenyl,4,4'-azodiphenetole,bis 4-ethoxyphenyl diazene,4,4'-azophenetole,bis-p-ethoxyazobenzene,e-bis 4-ethoxyphenyl diazene,diazene, 1,2-bis 4-ethoxyphenyl,phenetole, 4,4'-azodi-6ci,7ci,acmc-20amss PubChem CID: 101783 IUPAC Name: bis(4-ethoxyphenyl)diazene SMILES: CCOC1=CC=C(C=C1)N=NC1=CC=C(OCC)C=C1