missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diethoxyazobenzene, 97%

Catalog No. AAB2083903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2083903 1 g
AAB2083906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2083903 Supplier Thermo Scientific Chemicals Supplier No. B2083903

CAS: 588-52-3 | C16H18N2O2 | 270.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 588-52-3
Molecular Formula C16H18N2O2
Molecular Weight (g/mol) 270.33
MDL Number MFCD00053756
InChI Key FMPYMMTZQFFROU-UHFFFAOYSA-N
Synonym 4,4'-diethoxyazobenzene, diazene, bis 4-ethoxyphenyl, 4,4'-azodiphenetole, bis 4-ethoxyphenyl diazene, 4,4'-azophenetole, bis-p-ethoxyazobenzene, e-bis 4-ethoxyphenyl diazene, diazene, 1,2-bis 4-ethoxyphenyl, phenetole, 4,4'-azodi-6ci,7ci, acmc-20amss
PubChem CID 101783
IUPAC Name bis(4-ethoxyphenyl)diazene
SMILES CCOC1=CC=C(C=C1)N=NC1=CC=C(OCC)C=C1

Specifications

Quantity 1 g
Formula Weight 270.33
Percent Purity 97%
Chemical Name or Material 4,4'-Diethoxyazobenzene
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
RTECSNumber HM2966300
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only