missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorodiphenyl Sulfone 98.0+%, TCI America™

Catalog No. p-7118684
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B081025G 25 g
B0810100G 100 g
B0810500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B081025G Supplier TCI America Supplier No. B081025G
Only null left

Chemical Identifiers

CAS 80-07-9
Molecular Formula C12H8Cl2O2S
Molecular Weight (g/mol) 287.15
MDL Number MFCD00000619
InChI Key GPAPPPVRLPGFEQ-UHFFFAOYSA-N
Synonym 4,4'-dichlorodiphenyl sulfone, bis 4-chlorophenyl sulfone, p,p'-dichlorodiphenyl sulfone, sulfone, bis p-chlorophenyl, 4-chlorophenyl sulfone, 1,1'-sulfonylbis 4-chlorobenzene, bis p-chlorophenyl sulfone, benzene, 1,1'-sulfonylbis 4-chloro, p-chlorophenyl sulfone, 4,4'-dichlorodiphenylsulfone
PubChem CID 6625
IUPAC Name 1-chloro-4-(4-chlorobenzenesulfonyl)benzene
SMILES ClC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 148°C
Color White
Quantity 25 g
Formula Weight 287.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dichlorodiphenyl Sulfone
EINECSNumber (3)-0061&(3)-2160
RTECSNumber WR3450000
TSCA Yes