missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorodiphenyl sulfone, 97%

CAS: 80-07-9 | C12H8Cl2O2S | 287.15 g/mol

$101.98 - $101.98

Chemical Identifiers

CAS 80-07-9
Molecular Formula C12H8Cl2O2S
Molecular Weight (g/mol) 287.15
MDL Number MFCD00000619
InChI Key GPAPPPVRLPGFEQ-UHFFFAOYSA-N
Synonym 4,4'-dichlorodiphenyl sulfone, bis 4-chlorophenyl sulfone, p,p'-dichlorodiphenyl sulfone, sulfone, bis p-chlorophenyl, 4-chlorophenyl sulfone, 1,1'-sulfonylbis 4-chlorobenzene, bis p-chlorophenyl sulfone, benzene, 1,1'-sulfonylbis 4-chloro, p-chlorophenyl sulfone, 4,4'-dichlorodiphenylsulfone
PubChem CID 6625
IUPAC Name 1-chloro-4-(4-chlorophenyl)sulfonylbenzene
SMILES ClC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(Cl)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC291182500
SDS View Documents
Thermo Scientific Chemicals
291182500
250 g Plastic bottle
Each for $101.98
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

80-07-9
287.15
GPAPPPVRLPGFEQ-UHFFFAOYSA-N
6625
ClC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(Cl)C=C1
C12H8Cl2O2S
MFCD00000619
4,4'-dichlorodiphenyl sulfone, bis 4-chlorophenyl sulfone, p,p'-dichlorodiphenyl sulfone, sulfone, bis p-chlorophenyl, 4-chlorophenyl sulfone, 1,1'-sulfonylbis 4-chlorobenzene, bis p-chlorophenyl sulfone, benzene, 1,1'-sulfonylbis 4-chloro, p-chlorophenyl sulfone, 4,4'-dichlorodiphenylsulfone
1-chloro-4-(4-chlorophenyl)sulfonylbenzene

Specifications

80-07-9
100.0
White
233°C
96% min. (GC)
C12H8Cl2O2S
250 g
4,4'-dichlorodiphenyl sulfone, bis 4-chlorophenyl sulfone, p,p'-dichlorodiphenyl sulfone, sulfone, bis p-chlorophenyl, 4-chlorophenyl sulfone, 1,1'-sulfonylbis 4-chlorobenzene, bis p-chlorophenyl sulfone, benzene, 1,1'-sulfonylbis 4-chloro, p-chlorophenyl sulfone, 4,4'-dichlorodiphenylsulfone
GPAPPPVRLPGFEQ-UHFFFAOYSA-N
1-chloro-4-(4-chlorophenyl)sulfonylbenzene
6625
97%
4, 4'-Dichlorodiphenyl sulfone, 97%
96.0
146.0°C to 151.0°C
397.0°C
Authentic
Plastic bottle
MFCD00000619
06, 327
Solubility in water: .. Other solubilities: soluble in alcohols,slightly soluble in aqueous solvents
ClC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(Cl)C=C1
287.15
287.16
Fine Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause long lasting harmful effects to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 201-247-9

RTECSNumber : WR3450000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only