missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorodiphenyl Disulfide 98.0+%, TCI America™

Catalog No. D03605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D03605G 5 g
D036025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D03605G Supplier TCI America Supplier No. D03605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1142-19-4
Molecular Formula C12H8Cl2S2
Molecular Weight (g/mol) 287.216
MDL Number MFCD00013642
InChI Key ZIXXRXGPBFMPFD-UHFFFAOYSA-N
Synonym 4,4'-dichlorodiphenyl disulfide, bis 4-chlorophenyl disulfide, disulfide, bis 4-chlorophenyl, ddds, ddds pesticide, disulfide, bis p-chlorophenyl, p-chlorophenyl disulfide, 4-chlorophenyl disulfide, bis p-chlorophenyl disulfide, 4,4'-dichloro diphenyl disulfide
PubChem CID 14360
IUPAC Name 1-chloro-4-[(4-chlorophenyl)disulfanyl]benzene
SMILES C1=CC(=CC=C1SSC2=CC=C(C=C2)Cl)Cl

Specifications

Melting Point 72°C
Color Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Boiling Point 180°C
Formula Weight 287.22
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4,4′-Dichlorodiphenyl Disulfide
EINECSNumber (9)-0582
RTECSNumber JO0766000
TSCA Yes