missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diamino-2,2'-dimethylbibenzyl 95.0+%, TCI America™

Catalog No. D16585G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D16585G Supplier TCI America Supplier No. D16585G

Chemical Identifiers

CAS 54628-21-6
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD00007812
InChI Key ISESBQNCWCFFFR-UHFFFAOYSA-N
Synonym 4,4′C-Ethylenedi-m-toluidine
PubChem CID 89867
IUPAC Name 4-[2-(4-amino-2-methylphenyl)ethyl]-3-methylaniline
SMILES CC1=C(C=CC(=C1)N)CCC2=C(C=C(C=C2)N)C

Specifications

Melting Point 170°C
Quantity 5 g
Formula Weight 240.35
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diamino-2,2′-dimethylbibenzyl