missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dicarboxydiphenyl Sulfone 98.0+%, TCI America™
$441.78 - $441.78
Chemical Identifiers
| CAS | 2449-35-6 |
|---|---|
| Molecular Formula | C14H10O6S |
| Molecular Weight (g/mol) | 306.288 |
| MDL Number | MFCD00020375 |
| InChI Key | SQJQLYOMPSJVQS-UHFFFAOYSA-N |
| PubChem CID | 224167 |
| IUPAC Name | 4-(4-carboxyphenyl)sulfonylbenzoic acid |
| SMILES | C1=CC(=CC=C1C(=O)O)S(=O)(=O)C2=CC=C(C=C2)C(=O)O |
Chemical Identifiers
| 2449-35-6 | |
| 306.288 | |
| SQJQLYOMPSJVQS-UHFFFAOYSA-N | |
| 4-(4-carboxyphenyl)sulfonylbenzoic acid |
| C14H10O6S | |
| MFCD00020375 | |
| 224167 | |
| C1=CC(=CC=C1C(=O)O)S(=O)(=O)C2=CC=C(C=C2)C(=O)O |
Specifications
| 2449-35-6 | |
| White | |
| MFCD00020375 | |
| 4,4′C-Sulfonyldibenzoic Acid | |
| C1=CC(=CC=C1C(=O)O)S(=O)(=O)C2=CC=C(C=C2)C(=O)O | |
| 306.288 | |
| 306.29 | |
| Crystalline Powder |
| 370°C | |
| C14H10O6S | |
| 5 g | |
| SQJQLYOMPSJVQS-UHFFFAOYSA-N | |
| 4-(4-carboxyphenyl)sulfonylbenzoic acid | |
| 224167 | |
| ≥98.0% (HPLC) | |
| 4,4′-Dicarboxydiphenyl Sulfone |
Safety and Handling
TSCA : No