missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dicarboxydiphenyl Ether 98.0+%, TCI America™

Catalog No. p-7127115
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D211525G 25 g
D2115100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D211525G Supplier TCI America Supplier No. D211525G
Only null left

Chemical Identifiers

CAS 2215-89-6
Molecular Formula C14H10O5
Molecular Weight (g/mol) 258.229
MDL Number MFCD00013988
InChI Key WVDRSXGPQWNUBN-UHFFFAOYSA-N
Synonym 4,4'-oxybis benzoic acid, 4,4'-oxydibenzoic acid, 4,4'-oxybisbenzoic acid, 4-4-carboxyphenoxy benzoic acid, benzoic acid, 4,4'-oxybis, 4,4'-dicarboxydiphenyl ether, 4,4-oxobisbenzoic acid, 4,4'-diphenyl ether dicarboxylic acid, diphenyl ether 4,4'-dicarboxylic acid, pubchem9032
PubChem CID 75183
IUPAC Name 4-(4-carboxyphenoxy)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)OC2=CC=C(C=C2)C(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 258.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dicarboxydiphenyl Ether
EINECSNumber (3)-4453&(3)-4483
TSCA Yes